accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2COO

CAS RN 2705-87-5

IUPAC Name: prop-2-en-1-yl 3-cyclohexylpropanoate
Synonyms: allyl cyclohexylpropionate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: TWXUTZNBHUWMKJ-UHFFFAOYSA-N
SMILES: C=CCOC(=O)CCC1CCCCC1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)