accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOOOHOHOSOOOH2ONaNa

CAS RN 270573-71-2

IUPAC Name: disodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate
Synonyms: disodium 4,5-dihydroxybenzene-1,3-disulfonate hydrate
Molecular Weight (g/mol): 332.2
Molecular Formula: C6H6Na2O9S2
InChi Key: ZLRROLLKQDRDPI-UHFFFAOYSA-L
SMILES: O.[Na+].[Na+].OC1=CC(=CC(=C1O)S([O-])(=O)=O)S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (3)
  • (1)
Color 
  • (3)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)