accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHN(R)O(R)O(R)O(R)HO(S)HO(R)HO(R)NHH3CO(S)O(S)O(R)OH(S)OH(R)OH(R)NHCH3O(R)OHO(S)O(R)OH(S)OH(R)OH(R)HNCH3O

CAS RN 2706-65-2

IUPAC Name: N-[(2R,3R,4S,5R)-3-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-bis({[(2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})-6-hydroxy-1-oxohexan-2-yl]acetamide
Synonyms: N-[(2R,3R,4S,5R)-3-{[(2R,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-bis({[(2S,3R,4R,5S,6R)-3-acetamido-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy})-6-hydroxy-1-oxohexan-2-yl]acetamide
Molecular Weight (g/mol): 830.79
Molecular Formula: C32H54N4O21
InChi Key: QWBFSMSGJPCHBW-PNWIGLAMSA-N
SMILES: CC(=O)N[C@@H](C=O)[C@@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@H](O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O)[C@@H](CO)O[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1NC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results