accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2706-90-3

IUPAC Name: nonafluoropentanoic acid
Synonyms: nonafluoropentanoic acid
Molecular Weight (g/mol): 264.05
Molecular Formula: C5HF9O2
InChi Key: CXZGQIAOTKWCDB-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (6)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (1)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (7)
  • (2)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)