accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBFFFHC

CAS RN 27081-10-3

IUPAC Name: cyclohepta-2,4,6-trien-1-ylium; tetrafluoroboranuide
Synonyms: tropylium tetrafluoroborate
Molecular Weight (g/mol): 177.94
Molecular Formula: C7H7BF4
InChi Key: SQVQHTIKOZVGLR-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.[CH+]1C=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)