accessibility menu, dialog, popup

UserName

Viewing profile as user:
HNN

CAS RN 271-63-6

IUPAC Name: 1H-pyrrolo[2,3-b]pyridine
Synonyms: 7-azaindole
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: MVXVYAKCVDQRLW-UHFFFAOYSA-N
SMILES: N1C=CC2=CC=CN=C12
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (3)
  • (3)
  • (3)
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (7)
  • (3)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)