CAS RN 271-73-8
IUPAC Name:
1H-pyrazolo[3,4-b]pyridine
Synonyms:
1H-pyrazolo[3,4-b]pyridine
Molecular Weight (g/mol):
119.13
Molecular Formula:
C6H5N3
InChi Key:
GVLRTOYGRNLSDW-UHFFFAOYSA-N
SMILES:
N1N=CC2=CC=CN=C12