accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2713-31-7

IUPAC Name: 2,5-difluorophenol
Synonyms: 2,5-difluorophenol
Molecular Weight (g/mol): 130.09
Molecular Formula: C6H4F2O
InChi Key: INXKVYFOWNAVMU-UHFFFAOYSA-N
SMILES: OC1=CC(F)=CC=C1F
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)