accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27130-46-7

H3CNHOOHCl

CAS RN 27130-46-7

IUPAC Name: methylamino benzoate hydrochloride
Synonyms: methylamino benzoate hydrochloride
Molecular Weight (g/mol): 187.62
Molecular Formula: C8H10ClNO2
InChi Key: FTCDQNQIGUCFSA-UHFFFAOYSA-N
SMILES: Cl.CNOC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)