CAS RN 27174-15-8
IUPAC Name:
2-(2-aminoacetamido)-3-hydroxybutanoic acid
Synonyms:
2-(2-aminoacetamido)-3-hydroxybutanoic acid
Molecular Weight (g/mol):
176.17
Molecular Formula:
C6H12N2O4
InChi Key:
OLIFSFOFKGKIRH-UHFFFAOYNA-N
SMILES:
CC(O)C(NC(=O)CN)C(O)=O