accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27182-54-3

IUPAC Name: 5-phenyl-1,2,4-thiadiazol-3-amine
Synonyms: 5-phenyl-1,2,4-thiadiazol-3-amine
Molecular Weight (g/mol): 177.23
Molecular Formula: C8H7N3S
InChi Key: WOZDQVLZCXNJPY-UHFFFAOYSA-N
SMILES: NC1=NSC(=N1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results