accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNNN

CAS RN 27187-13-9

IUPAC Name: 5-chloro-[1,2,4]triazolo[4,3-a]pyridine
Synonyms: 5-chloro-[1,2,4]triazolo[4,3-a]pyridine
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: FBQZXTMUYNKLRF-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=NN=CN12
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)