accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2719-23-5

IUPAC Name: N-(1,3-thiazol-2-yl)acetamide
Synonyms: acetamide, N-2-thiazolyl-
Molecular Weight (g/mol): 142.18
Molecular Formula: C5H6N2OS
InChi Key: WXPLRSVMGRAIGW-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results