accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2719-30-4

IUPAC Name: 1-isothiocyanato-2-nitrobenzene
Synonyms: 1-isothiocyanato-2-nitrobenzene
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: CBWJHIXSVFDERH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1N=C=S
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results