CAS RN 27192-91-2
IUPAC Name:
2,2',7,7'-tetrabromo-9,9'-bifluorenylidene
Synonyms:
2,2',7,7'-tetrabromo-9,9'-bifluorenylidene
Molecular Weight (g/mol):
644
Molecular Formula:
C26H12Br4
InChi Key:
ZPFZLDRAZNLKJI-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)C2=C1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
1
–
1
of
1
results
1
2,2',7,7'-Tetrabromo-9,9'-bifluorenylidene 98.0+%, TCI America™
CAS: 27192-91-2 Molecular Formula: C26H12Br4 Molecular Weight (g/mol): 643.998 InChI Key: ZPFZLDRAZNLKJI-UHFFFAOYSA-N PubChem CID: 13806537 IUPAC Name: 2,7-dibromo-9-(2,7-dibromofluoren-9-ylidene)fluorene SMILES: C1=CC2=C(C=C1Br)C(=C3C4=C(C=CC(=C4)Br)C5=C3C=C(C=C5)Br)C6=C2C=CC(=C6)Br