accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27208-80-6

IUPAC Name: (2S,3R,4S,5S,6R)-2-{3-hydroxy-5-[(1E)-2-(4-hydroxyphenyl)ethenyl]phenoxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: piceid polydatin
Molecular Weight (g/mol): 390.39
Molecular Formula: C20H22O8
InChi Key: HSTZMXCBWJGKHG-CUYWLFDKSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC(O)=CC(\C=C\C3=CC=C(O)C=C3)=C2)[C@H](O)[C@@H](O)[C@@H]1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results