accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNSHN

CAS RN 27231-36-3

IUPAC Name: 5-methyl-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 2-mercaptomethylbenzimidazole
Molecular Weight (g/mol): 164.23
Molecular Formula: C8H8N2S
InChi Key: CWIYBOJLSWJGKV-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=S)NC2=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)