CAS RN 2724-69-8
CAS RN 2724-69-8
IUPAC Name:
3-methyl-1-phenylthiourea
Synonyms:
1-methyl-3-phenylthiourea
Molecular Weight (g/mol):
166.24
Molecular Formula:
C8H10N2S
InChi Key:
IGEQFPWPMCIYDF-UHFFFAOYSA-N
SMILES:
CNC(=S)NC1=CC=CC=C1
1
–
1
of
1
results
1
1-Methyl-3-phenylthiourea 97.0+%, TCI America™
CAS: 2724-69-8 Molecular Formula: C8H10N2S Molecular Weight (g/mol): 166.242 MDL Number: MFCD00022130 InChI Key: IGEQFPWPMCIYDF-UHFFFAOYSA-N PubChem CID: 698294 IUPAC Name: 1-methyl-3-phenylthiourea SMILES: CNC(=S)NC1=CC=CC=C1