accessibility menu, dialog, popup

UserName

Viewing profile as user:
ZnCH3H3CCH3H3CCH3OOCH3H3CCH3H3CCH3OO

CAS RN 27253-29-8

IUPAC Name: zinc(2+) bis(3,3,5,5-tetramethylhexanoate)
Synonyms: zinc neodecanoate
Molecular Weight (g/mol): 407.9
Molecular Formula: C20H38O4Zn
InChi Key: JUODUXKPGONLIM-UHFFFAOYSA-L
SMILES: [Zn++].CC(C)(C)CC(C)(C)CC([O-])=O.CC(C)(C)CC(C)(C)CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)