accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 273-21-2

IUPAC Name: 1H-imidazo[4,5-b]pyridine
Synonyms: 3H-imidazo[4,5-b]pyridine 1-deazapurine 7-azabenzimidazole azabenzimidazole
Molecular Weight (g/mol): 119.13
Molecular Formula: C6H5N3
InChi Key: GAMYYCRTACQSBR-UHFFFAOYSA-N
SMILES: N1C=NC2=NC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)