accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHClONNFFF

CAS RN 27314-13-2

IUPAC Name: 4-chloro-5-(methylamino)-2-[3-(trifluoromethyl)phenyl]-2,3-dihydropyridazin-3-one
Synonyms: telok zorial solicam rapid zorial rapid 80 monometflurazone solicam caswell no. 195AA evital evitol C12H9clF3N3O
Molecular Weight (g/mol): 303.67
Molecular Formula: C12H9ClF3N3O
InChi Key: NVGOPFQZYCNLDU-UHFFFAOYSA-N
SMILES: CNC1=C(Cl)C(=O)N(N=C1)C1=CC=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results