accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 273221-75-3

IOOHOOHN(S)CH3H3CCH3

CAS RN 273221-75-3

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(3-iodophenyl)propanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-3-(3-iodophenyl)propanoic acid
Molecular Weight (g/mol): 391.21
Molecular Formula: C14H18INO4
InChi Key: MPTBEGFNTNOLNZ-NSHDSACASA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC(I)=CC=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)