accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27344-41-8

IUPAC Name: disodium 2-[(1Z)-2-{4'-[(1Z)-2-(2-sulfonatophenyl)ethenyl]-[1,1'-biphenyl]-4-yl}ethenyl]benzene-1-sulfonate
Synonyms: disodium distyrylbiphenyl disulfonate
Molecular Weight (g/mol): 562.56
Molecular Formula: C28H20Na2O6S2
InChi Key: PMPJQLCPEQFEJW-GNTLFSRWSA-L
SMILES: [Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1\C=C/C1=CC=C(C=C1)C1=CC=C(\C=C/C2=CC=CC=C2S([O-])(=O)=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results