accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27347-32-6

IUPAC Name: 2,3-dihydro-1-benzofuran-2-carbonyl chloride
Synonyms: 2,3-dihydro-1-benzofuran-2-carbonyl chloride
Molecular Weight (g/mol): 182.6
Molecular Formula: C9H7ClO2
InChi Key: QZVPHVMVWZKULP-UHFFFAOYNA-N
SMILES: ClC(=O)C1CC2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results