accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2736-40-5

IUPAC Name: 2-ethylbutanoyl chloride
Synonyms: butanoyl chloride, 2-ethyl-
Molecular Weight (g/mol): 134.6
Molecular Formula: C6H11ClO
InChi Key: SMUKODJVMQOSAB-UHFFFAOYSA-N
SMILES: CCC(CC)C(Cl)=O
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)