accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2737-22-6

IUPAC Name: 2,4,6-tribromo-3-hydroxybenzaldehyde
Synonyms: 2,4,6-tribromo-3-hydroxybenzaldehyde
Molecular Weight (g/mol): 358.81
Molecular Formula: C7H3Br3O2
InChi Key: FAWOIFAFFUDNJX-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(Br)C(C=O)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results