accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOSiH3CCH3H3C

CAS RN 27374-25-0

IUPAC Name: (1-ethoxycyclopropoxy)trimethylsilane
Synonyms: (1-ethoxycyclopropoxy)trimethylsilane
Molecular Weight (g/mol): 174.32
Molecular Formula: C8H18O2Si
InChi Key: BZMMRNKDONDVIB-UHFFFAOYSA-N
SMILES: CCOC1(CC1)O[Si](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)