accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2738-19-4

IUPAC Name: 2-methylhex-2-ene
Synonyms: 2-methyl-2-hexene
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: BWEKDYGHDCHWEN-UHFFFAOYSA-N
SMILES: CCCC=C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results