accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOONOO

CAS RN 27402-68-2

IUPAC Name: 3',6'-dihydroxy-6-nitro-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 3',6'-dihydroxy-6-nitrospiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 377.31
Molecular Formula: C20H11NO7
InChi Key: GYWGQAIDMYGOLU-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC=C(C=C23)[N+]([O-])=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results