accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2CP

CAS RN 2741-38-0

IUPAC Name: diphenyl(prop-2-en-1-yl)phosphane
Synonyms: diphenyl(prop-2-en-1-yl)phosphane
Molecular Weight (g/mol): 226.26
Molecular Formula: C15H15P
InChi Key: PDDYFPPQDKRJTK-UHFFFAOYSA-N
SMILES: C=CCP(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)