accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OOHNN

CAS RN 27425-55-4

IUPAC Name: 3-(1H-1,3-benzodiazol-2-yl)-7-(diethylamino)-2H-chromen-2-one
Synonyms: 3-(1H-1,3-benzodiazol-2-yl)-7-(diethylamino)chromen-2-one
Molecular Weight (g/mol): 333.39
Molecular Formula: C20H19N3O2
InChi Key: GOLORTLGFDVFDW-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C2C=C(C3=NC4=CC=CC=C4N3)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results