accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27428-57-5

OBrCl

CAS RN 27428-57-5

IUPAC Name: (4-bromophenyl)(4-chlorophenyl)methanone
Synonyms: (4-bromophenyl)(4-chlorophenyl)methanone
Molecular Weight (g/mol): 295.56
Molecular Formula: C13H8BrClO
InChi Key: FYMCXGILCMIOKD-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results