accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27431-62-5

IUPAC Name: (4-aminobutyl)diethylamine
Synonyms: (4-aminobutyl)diethylamine
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: JILXUIANNUALRZ-UHFFFAOYSA-N
SMILES: CCN(CC)CCCCN
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)