accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CCH3Br

CAS RN 27452-17-1

IUPAC Name: 6-bromo-1,1,4,4-tetramethyl-1,2,3,4-tetrahydronaphthalene
Synonyms: 6-bromo-1,1,4,4-tetramethyl-2,3-dihydronaphthalene
Molecular Weight (g/mol): 267.21
Molecular Formula: C14H19Br
InChi Key: NLOOVMVNNNYLFS-UHFFFAOYSA-N
SMILES: CC1(C)CCC(C)(C)C2=CC(Br)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results