accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SCH3NN

CAS RN 27464-82-0

IUPAC Name: dimethyl-1,3,4-thiadiazole
Synonyms: dimethyl-1,3,4-thiadiazole
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: JXQGICFGPUAVLJ-UHFFFAOYSA-N
SMILES: CC1=NN=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results