accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27466-83-7

IUPAC Name: 4-bromo-N-methylbenzamide
Synonyms: 4-bromo-N-methylbenzamide
Molecular Weight (g/mol): 214.06
Molecular Formula: C8H8BrNO
InChi Key: DXCFWNVWQTYPOC-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results