accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNHNH2F

CAS RN 27469-60-9

IUPAC Name: 4-[bis(4-fluorophenyl)methyl]piperazine-1,4-diium
Synonyms: 4-[bis(4-fluorophenyl)methyl]piperazine-1,4-diium
Molecular Weight (g/mol): 290.36
Molecular Formula: C17H20F2N2
InChi Key: TTXIFFYPVGWLSE-UHFFFAOYSA-P
SMILES: FC1=CC=C(C=C1)C([NH+]1CC[NH2+]CC1)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)