accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27485-15-0

IUPAC Name: 9,10-dioxo-9,10-dihydroanthracene-2,3-dicarboxylic acid
Synonyms: 9,10-dioxoanthracene-2,3-dicarboxylic acid
Molecular Weight (g/mol): 296.23
Molecular Formula: C16H8O6
InChi Key: PSHVQIKBLXBIQJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C2C(=O)C3=CC=CC=C3C(=O)C2=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results