accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNCH3O

CAS RN 2749-59-9

IUPAC Name: 1,3-dimethyl-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 2,5-dimethyl-4H-pyrazol-3-one
Molecular Weight (g/mol): 112.13
Molecular Formula: C5H8N2O
InChi Key: NDELSWXIAJLWOU-UHFFFAOYSA-N
SMILES: CN1N=C(C)CC1=O
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)