accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHONHOOCH3CH3H3CHN

CAS RN 27494-47-9

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}butanoic acid; N-cyclohexylcyclohexanamine
Synonyms: 2-[(tert-butoxycarbonyl)amino]butanoic acid; dicha
Molecular Weight (g/mol): 384.56
Molecular Formula: C21H40N2O4
InChi Key: HOSMYZXDFVUSCV-UHFFFAOYNA-N
SMILES: C1CCC(CC1)NC1CCCCC1.CCC(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results