CAS RN 27496-82-8
IUPAC Name:
3-[(3-carboxy-2-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
Synonyms:
3-[(3-carboxy-2-hydroxyphenyl)methyl]-2-hydroxybenzoic acid
Molecular Weight (g/mol):
288.26
Molecular Formula:
C15H12O6
InChi Key:
RWXFMUMBJLQGDZ-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=CC(CC2=CC=CC(C(O)=O)=C2O)=C1O
1
–
1
of
1
results
1
Methylenedisalicylic Acid (mixture of isomers) 90.0+%, TCI America™
CAS: 27496-82-8 Molecular Formula: C15H12O6 MDL Number: MFCD00016506