accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrP

CAS RN 2751-90-8

IUPAC Name: tetraphenylphosphanium bromide
Synonyms: tetraphenylphosphonium bromide
Molecular Weight (g/mol): 419.3
Molecular Formula: C24H20BrP
InChi Key: BRKFQVAOMSWFDU-UHFFFAOYSA-M
SMILES: [Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (5)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (2)
  • (2)
  • (6)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (8)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)