accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27519-02-4

IUPAC Name: (9Z)-tricos-9-ene
Synonyms: (Z)-9-tricosene
Molecular Weight (g/mol): 322.62
Molecular Formula: C23H46
InChi Key: IGOWHGRNPLFNDJ-ZPHPHTNESA-N
SMILES: CCCCCCCCCCCCC\C=C/CCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results