accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27527-05-5

IUPAC Name: 2-amino-3-cyclohexylpropanoic acid
Synonyms: cyclohexylalanine
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: ORQXBVXKBGUSBA-UHFFFAOYNA-N
SMILES: NC(CC1CCCCC1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)