accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27532-96-3

IUPAC Name: 2-(tert-butoxy)-2-oxoethan-1-aminium
Synonyms: 2-(tert-butoxy)-2-oxoethanaminium
Molecular Weight (g/mol): 132.18
Molecular Formula: C6H14NO2
InChi Key: SJMDMGHPMLKLHQ-UHFFFAOYSA-O
SMILES: CC(C)(C)OC(=O)C[NH3+]
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (4)