accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27538-10-9

IUPAC Name: 2-ethyl-4-hydroxy-5-methyl-2,3-dihydrofuran-3-one
Synonyms: 2-ethyl-4-hydroxy-5-methyl-2H-furan-3-one
Molecular Weight (g/mol): 142.15
Molecular Formula: C7H10O3
InChi Key: GWCRPYGYVRXVLI-UHFFFAOYNA-N
SMILES: CCC1OC(C)=C(O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results