accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2754-27-0

IUPAC Name: trimethylsilyl acetate
Synonyms: trimethylsilyl acetate
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12O2Si
InChi Key: QHUNJMXHQHHWQP-UHFFFAOYSA-N
SMILES: CC(=O)O[Si](C)(C)C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)