accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2756-85-6

IUPAC Name: 1-aminocyclohexane-1-carboxylic acid
Synonyms: 1-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: WOXWUZCRWJWTRT-UHFFFAOYSA-N
SMILES: NC1(CCCCC1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)