accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NHOOHNH2N

CAS RN 2760-98-7

IUPAC Name: benzene-1,3-dicarbohydrazide
Synonyms: benzene-1,3-dicarbohydrazide
Molecular Weight (g/mol): 194.19
Molecular Formula: C8H10N4O2
InChi Key: UTTHLMXOSUFZCQ-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC(=CC=C1)C(=O)NN
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results