accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHONH2(E/Z)

CAS RN 27620-10-6

IUPAC Name: N'-hydroxybutanimidamide
Synonyms: N'-hydroxybutanimidamide
Molecular Weight (g/mol): 102.14
Molecular Formula: C4H10N2O
InChi Key: OPENCMFJZQABIY-UHFFFAOYSA-N
SMILES: CCCC(N)=NO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results